Proteintech
Manufacturer Code:210761AP150UL
Catalog # 50-173-3245
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | C1orf107 Fusion Protein Ag15250 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C1orf107 DEF; Digestive organ expansion factor homolog; digestive-organ expansion factor homolog
Gene Aliases: C1orf107; DEF; DIEXF; DJ434O14.5; UTP25
UniProt ID: (Human) Q68CQ4
Entrez Gene ID: (Human) 27042
Molecular Function:
RNA binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.