Proteintech
Manufacturer Code:215061AP150UL
Catalog # 50-173-3454
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | C1orf106 Fusion Protein Ag15987 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C1orf106; Innate immunity activator protein; uncharacterized protein C1orf106 homolog
Gene Aliases: 1700034M08Rik; 4933426C09Rik; AI586180; C1orf106; D1Mgi54; INAVA
UniProt ID: (Mouse) Q7TN12, (Human) Q3KP66
Entrez Gene ID: (Mouse) 67313, (Human) 55765, (Rat) 289399
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.