Abnova
Manufacturer Code:PAB13152
Catalog # 89-116-400
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to N-terminus 15 amino acids of human C1QTNF1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of C1QTNF1.
For Research Use Only
Protein Aliases: Complement C1q tumor necrosis factor-related protein 1; CTRP1; FLJ90694; G protein coupled receptor interacting protein; G protein-coupled receptor-interacting protein; GIP; ZSIG37
Gene Aliases: C1QTNF1; CTRP1; GIP; UNQ310/PRO353; ZSIG37
UniProt ID: (Human) Q9BXJ1
Entrez Gene ID: (Human) 114897
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.