Invitrogen
Manufacturer Code:PA581322
Catalog # PIPA581322
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Mouse C1QBP/HABP1 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Complement component 1 Q subcomponent-binding protein mitochondrial &lsqbPrecursor] Glycoprotein gC1qBP C1qBP GC1q-R protein Hyaluro-binding protein 1 Mitochondrial matrix protein p32 p33 C1QBP GC1QBP HABP1 SF2P32; complement component 1 Q subcomponent-binding protein, mitochondrial; GC1q-R protein; glycoprotein gC1qBP
Gene Aliases: AA407365; AA986492; D11Wsu182e; gC1qBP; HABP1; P32
Entrez Gene ID: (Mouse) 12261
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.