Invitrogen
Manufacturer Code:PA555318
Catalog # PIPA555318
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human C1QBP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ASF/SF2-associated protein p32; C1q globular domain-binding protein; C1qBP; Complement component 1 Q subcomponent-binding protein mitochondrial &lsqbPrecursor] Glycoprotein gC1qBP C1qBP GC1q-R protein Hyaluro-binding protein 1 Mitochondrial matrix protein p32 p33 C1QBP GC1QBP HABP1 SF2P32; Complement component 1 Q subcomponent-binding protein, mitochondrial; complement component 1, q subcomponent binding protein; GC1q-R protein; Glycoprotein gC1qBP; Hyaluronan-binding protein 1; Mitochondrial matrix protein p32; p33; SF2AP32; splicing factor SF2-associated protein
Gene Aliases: AA407365; AA986492; C1QBP; D11Wsu182e; gC1Q-R; GC1QBP; gC1qR; HABP1; P32; SF2P32
UniProt ID: (Human) Q07021, (Rat) O35796
Entrez Gene ID: (Human) 708, (Rat) 29681, (Mouse) 12261
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.