Invitrogen
Manufacturer Code:PA576228
Catalog # PIPA576228
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein of human C1GALT1C1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: beta 1,3-galactosyltransferase 2; beta1,3-galactosyltransferase 2; C1Gal-T2; C1GALT1-specific chaperone 1; C38H2-L1; C38H2-like protein 1; Core 1 beta1,3-galactosyltransferase 2; core 1 beta3-Gal-T2; Core 1 beta3-galactosyltransferase-specific molecular chaperone; core 1 UDP-galactose:N-acetylgalactosamine-alpha-R beta 1,3-galactosyltransferase 2; mC1Gal-T2; TNPS COSMC MST143 C1GALT2 HSPC067 C38H2-L1
Gene Aliases: 1500002I11Rik; AB030184; C1GALT1C1; C1GALT2; C38H2-L1; C81205; COSMC; HSPC067; MST143; MSTP143; RGD1311230; TNPS; UNQ273/PRO310
UniProt ID: (Human) Q96EU7, (Mouse) Q9JMG2, (Rat) Q499P3
Entrez Gene ID: (Human) 29071, (Mouse) 59048, (Rat) 302499
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.