Invitrogen
Manufacturer Code:PA5112371
Catalog # PIPA5112371
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human C1GALT1-specific chaperone 1 protein (30-190AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1500002I11Rik AB030184 beta 13-galactosyltransferase 2 beta13-galactosyltransferase 2 BOS_25149 C1GALT1 specific chaperone 1 C1GALT1C1 C1GALT1-specific chaperone 1 C1GALT1-specific chaperone 1-like C1GALT2 C1Gal-T2 C38H2-L1 C38H2-like protein 1 C81205 Core 1 beta13-galactosyltransferase 2 Core 1 beta3-galactosyltransferase-specific molecular chaperone core 1 beta3-Gal-T2 core 1 UDP-galactose:N-acetylgalactosamine-alpha-R beta 13-galactosyltransferase 2 Cosmc HSPC067 LOC100340186 mC1Gal-T2 MST143 MSTP143 RGD1311230 TNPS UNQ273/PRO310; beta 1,3-galactosyltransferase 2; beta1,3-galactosyltransferase 2; C1Gal-T2; C1GALT1-specific chaperone 1; C38H2-L1; C38H2-like protein 1; Core 1 beta1,3-galactosyltransferase 2; core 1 beta3-Gal-T2; Core 1 beta3-galactosyltransferase-specific molecular chaperone; mC1Gal-T2
Gene Aliases: 1500002I11Rik; AB030184; C1GALT1C1; C1GALT2; C38H2-L1; C81205; COSMC; HSPC067; MST143; MSTP143; TNPS; UNQ273/PRO310
UniProt ID: (Human) Q96EU7, (Mouse) Q9JMG2
Entrez Gene ID: (Human) 29071, (Mouse) 59048
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.