Novus Biologicals
Manufacturer Code:NBP2845450.1ML
Catalog # NB032473
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the N-terminal region of Rat C1GALT1C1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: beta 1,3-galactosyltransferase 2; beta 13-galactosyltransferase 2 C1GALT1-specific chaperone 1 C1GalT2 C1Gal-T2 C1GALT2MGC19947 C38H2-L1 Core 1 beta13-galactosyltransferase 2 Core 1 beta3-galactosyltransferase-specific molecular chaperone Core 1 beta3-Gal-T2 COSMCC38H2-like protein 1 MST143; C1Gal-T2; C1GALT1-specific chaperone 1; C38H2-L1; C38H2-like protein 1; Core 1 beta1,3-galactosyltransferase 2; core 1 beta3-Gal-T2; Core 1 beta3-galactosyltransferase-specific molecular chaperone
Gene Aliases: C1GALT1C1; C1GALT2; C38H2-L1; COSMC; HSPC067; MST143; MSTP143; TNPS; UNQ273/PRO310
UniProt ID: (Human) Q96EU7
Entrez Gene ID: (Human) 29071
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.