Abnova
Manufacturer Code:H00029071B01P
Catalog # 89-017-062
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | C1GALT1C1 (NP_001011551.1 1 a.a. ~ 318 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human C1GALT1C1 protein.
For Research Use Only
Protein Aliases: beta 1,3-galactosyltransferase 2; C1Gal-T2; C1GALT1-specific chaperone 1; C1GALT2; C38H2-L1; C38H2-like protein 1; Core 1 beta1,3-galactosyltransferase 2; core 1 beta3-Gal-T2; Core 1 beta3-galactosyltransferase-specific molecular chaperone; COSMC; HSPC067; MGC19947
Gene Aliases: C1GALT1C1; C1GALT2; C38H2-L1; COSMC; HSPC067; MST143; MSTP143; TNPS; UNQ273/PRO310
UniProt ID: (Human) Q96EU7
Entrez Gene ID: (Human) 29071
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.