Invitrogen
Manufacturer Code:PA543363
Catalog # PIPA543363
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human C1GALT1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B3Gal-T8; Beta-1,3-galactosyltransferase; C1GalT1; Core 1 beta1,3-galactosyltransferase 1; core 1 beta3-Gal-T; core 1 beta3-Gal-T1; Core 1 O-glycan T-synthase; Core 1 synthase glycoprotein-N-acetylgalactosamine 3-beta-galactosyltransferase 1 C1GALT T-synthase; core 1 synthase, glycoprotein-N-acetylgalactosamine 3-beta-galactosyltransferase, 1; Core 1 UDP-galactose:N-acetylgalactosamine-alpha-R beta 1,3-galactosyltransferase 1; core1 UDP-galactose:N-acetylgalactosamine-alpha-R beta 1,3-galactosyltransferase (C1galt1); core1 UDP-galactose:N-acetylgalactosamine-alpha-R beta 1,3-galactosyltransferase 1; core1 UDP-galactose:N-acetylgalactosamine-alpha-R beta 13-galactosyltransferase (C1galt1); Glycoprotein-N-acetylgalactosamine 3-beta-galactosyltransferase 1; T-syn
Gene Aliases: 2210410E06Rik; AV284120; C1GALT; C1GALT1; Plt1; T-synthase
UniProt ID: (Bovine) Q0VC84, (Human) Q9NS00, (Rat) Q9JJ05, (Mouse) Q9JJ06
Entrez Gene ID: (Horse) 100063865, (Bovine) 539417, (Dog) 482314, (Human) 56913, (Rabbit) 100359047, (Rat) 65044, (Mouse) 94192, (Guinea pig) 100714236
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.