Proteintech
Manufacturer Code:255141AP150UL
Catalog # 50-173-4941
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | C19orf70 Fusion Protein Ag22003 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C19orf70 MIC13 P117 Protein P117 Protein QIL1 QIL1; MICOS complex subunit MIC13; Protein P117; protein QIL1
Gene Aliases: C19orf70; MIC13; MICOS13; P117; QIL1; sr104
UniProt ID: (Mouse) Q8R404, (Human) Q5XKP0
Entrez Gene ID: (Mouse) 224904, (Human) 125988
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.