Proteintech
Manufacturer Code:194201AP150UL
Catalog # 50-173-2395
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | C19orf43 Fusion Protein Ag13747 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C19orf43 fSAP18; Exoribonuclease TRIR; My029 protein; Telomerase RNA component interacting RNase; uncharacterized protein C19orf43 homolog
Gene Aliases: 1110002H14Rik; AI854688; C19orf43; fSAP18; TRIR
UniProt ID: (Mouse) Q9D735, (Human) Q9BQ61
Entrez Gene ID: (Mouse) 68544, (Human) 79002, (Rat) 288920
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.