OriGene
Manufacturer Code:TA808727
Catalog # 50-168-2330
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI6F7 |
Immunogen | Human recombit protein fragment corresponding to amino acids 22-198 of human C19ORF80 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: angiopoietin-like 8; Angiopoietin-like protein 8; Betatrophin; betatrophin variant 1; betatrophin variant 2; C19orf80 PRO1185 PVPA599 RIFL TD26; hepatocellular carcinoma-associated gene TD26; hepatocellular carcinoma-associated protein TD26; Lipasin; Refeeding-induced fat and liver protein
Gene Aliases: ANGPTL8; C19orf80; PRO1185; PVPA599; RIFL; TD26; UNQ599/PRO1185
UniProt ID: (Human) Q6UXH0
Entrez Gene ID: (Human) 55908
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.