Proteintech
Manufacturer Code:201111AP150UL
Catalog # 50-173-2705
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | C18orf8 Fusion Protein Ag13904 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C18orf8 HsT2591 Mic 1 MIC1; Colon cancer-associated protein Mic1; colon cancer-associated protein Mic1 homolog; macrophage inhibitory cytokine 1; Mic-1; Regulator of MON1-CCZ1 complex; uncharacterized protein C18orf8 homolog; WD repeat-containing protein 98
Gene Aliases: 2400010D15Rik; C18orf8; HsT2591; Mic-1; MIC1; RMC1; WDR98
UniProt ID: (Mouse) Q8VC42, (Human) Q96DM3
Entrez Gene ID: (Mouse) 76482, (Human) 29919, (Rat) 291784
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.