Proteintech
Manufacturer Code:166411AP150UL
Catalog # 50-173-1078
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | C18orf55 Fusion Protein Ag10002 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C18orf55 HSPC154 Mitochondrial import inner membrane translocase subunit Tim21 TIM21 TIMM21; Mitochondrial import inner membrane translocase subunit Tim21; TIM21-like protein, mitochondrial; translocase of inner mitochondrial membrane 21 homolog
Gene Aliases: C18orf55; HSPC154; TIM21; TIMM21
UniProt ID: (Human) Q9BVV7
Entrez Gene ID: (Human) 29090
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.