OriGene
Manufacturer Code:TA800020
Catalog # 50-167-8009
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2D5 |
Immunogen | Full length human recombit protein of human C17orf37 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: C17orf37 C35 ORB3 RDX12 XTP4; HBV X-transactivated gene 4 protein; HBV XAg-transactivated protein 4; Migration and invasion enhancer 1; protein C17orf37; Protein C35
Gene Aliases: C17orf37; C35; MIEN1; ORB3; RDX12; XTP4
UniProt ID: (Human) Q9BRT3
Entrez Gene ID: (Human) 84299
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.