OriGene
Manufacturer Code:CF811338
Catalog # 50-167-0601
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4B1 |
Immunogen | Full length human recombit protein of human C16orf57 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: C16orf57 HVSL1 Mpn1 PN hUsb1; hUsb1; HVSL motif containing 1; mutated in poikiloderma with neutropenia protein 1; putative U6 snRNA phosphodiesterase; U six biogenesis 1; U6 snRNA biogenesis 1; U6 snRNA phosphodiesterase; UPF0406 protein C16orf57; USB1
Gene Aliases: C16orf57; hUsb1; HVSL1; Mpn1; PN; USB1
UniProt ID: (Human) Q9H8X8
Entrez Gene ID: (Human) 79650
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.