Invitrogen
Manufacturer Code:PA537975
Catalog # PIPA537975
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-TANQVKIYTNQEKT from the internal region of C16orf57 (aa154-167) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: chromosome 16 open reading frame 57 FLJ13154 HVSL1 PN HVSL motif containing 1 hypothetical protein LOC79650; hUsb1; HVSL motif containing 1; mutated in poikiloderma with neutropenia protein 1; putative U6 snRNA phosphodiesterase; U six biogenesis 1; U6 snRNA biogenesis 1; U6 snRNA phosphodiesterase; U6 snRNA phosphodiesterase {ECO:0000255|HAMAP-Rule:MF_03040}; UPF0406 protein C16orf57; UPF0406 protein C16orf57 homolog; USB1
Gene Aliases: AA960436; C16orf57; hUsb1; HVSL1; Mpn1; PN; RGD1305215; USB1
UniProt ID: (Human) Q9H8X8, (Mouse) Q91W78, (Rat) Q5I0I5
Entrez Gene ID: (Human) 79650, (Mouse) 101985, (Rat) 307643
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.