Abnova
Manufacturer Code:PAB24559
Catalog # 89-126-275
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 154-167 at internal region of human C16orf57. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of C16orf57.
For Research Use Only
Protein Aliases: FLJ13154; hUsb1; HVSL motif containing 1; HVSL1; mutated in poikiloderma with neutropenia protein 1; putative U6 snRNA phosphodiesterase; U six biogenesis 1; U6 snRNA biogenesis 1; U6 snRNA phosphodiesterase; UPF0406 protein C16orf57; USB1
Gene Aliases: C16orf57; hUsb1; HVSL1; Mpn1; PN; USB1
UniProt ID: (Human) Q9H8X8
Entrez Gene ID: (Human) 79650
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.