Proteintech
Manufacturer Code:208081AP150UL
Catalog # 50-173-3100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | C16orf14 Fusion Protein Ag14754 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C16orf14 c349E10.1 FAM195A Protein FAM195A; family with sequence similarity 195 member A; family with sequence similarity 195, member A; MAPK regulated co-repressor interacting protein 2; MAPK regulated corepressor interacting protein 2; Protein FAM195A
Gene Aliases: 9530058B02Rik; C16orf14; c349E10.1; FAM195A; MCRIP2
UniProt ID: (Mouse) Q9CQB2, (Human) Q9BUT9
Entrez Gene ID: (Mouse) 68241, (Human) 84331
Molecular Function:
kinase
protein kinase
protein kinase receptor
receptor
transferase
tyrosine protein kinase receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.