Proteintech
Manufacturer Code:208011AP150UL
Catalog # 50-173-3093
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | C16orf13 Fusion Protein Ag14718 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C16orf13 JFP2 UPF0585 protein C16orf13; Methyltransferase-like 26; UPF0585 protein C16orf13 homolog
Gene Aliases: C16orf13; JFP2; METTL26
UniProt ID: (Mouse) Q9DCS2, (Human) Q96S19, (Rat) Q497C3
Entrez Gene ID: (Mouse) 68347, (Human) 84326, (Rat) 302998
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.