Invitrogen
Manufacturer Code:PA525287
Catalog # PIPA525287
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 145-171 amino acids from the central region of human C14orf179 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C14orf179 CED3 IFT complex A subunit intraflagellar transport protein 43 homolog; IFT complex A subunit; intraflagellar transport 43 homolog; Intraflagellar transport protein 43 homolog
Gene Aliases: 1700019E19Rik; C14orf179; CED3; IFT43; R75064
UniProt ID: (Human) Q96FT9, (Mouse) Q9DA69
Entrez Gene ID: (Human) 112752, (Mouse) 76411
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.