Proteintech
Manufacturer Code:247221AP150UL
Catalog # 50-173-4506
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | C14orf138 Fusion Protein Ag20154 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C14orf138; methyltransferase like 21D; Methyltransferase-like protein 21D; Protein-lysine methyltransferase METTL21D; valosin containing protein lysine (K) methyltransferase; Valosin-containing protein lysine methyltransferase; VCP lysine methyltransferase; VCP-KMT
Gene Aliases: C14orf138; METTL21D; VCP-KMT; VCPKMT
UniProt ID: (Human) Q9H867
Entrez Gene ID: (Human) 79609
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.