Abnova
Manufacturer Code:H00051528K
Catalog # 89-204-378
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human C14ORF100 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human C14orf100 peptide using ARM Technology.
For Research Use Only
Protein Aliases: CDA06 HSPC213 HSPC327 JAMP; JAMP; JKAMP; JNK-associated membrane protein; JNK1-associated membrane protein; JNK1/MAPK8-associated membrane protein; Jun N-terminal kinase 1-associated membrane protein; Medulloblastoma antigen MU-MB-50.4
Gene Aliases: C14orf100; CDA06; HSPC213; HSPC327; JAMP; JKAMP
UniProt ID: (Human) Q9P055
Entrez Gene ID: (Human) 51528
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.