Invitrogen
Manufacturer Code:PA566622
Catalog # PIPA566622
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human C13orf15 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Regulator of cell cycle RGCC; Response gene to complement 32 protein; RGC-32; RGC-32 RGC32 bA157L14.2 response gene to complement 32 protein
Gene Aliases: bA157L14.2; C13orf15; RGC-32; RGC32; RGCC
UniProt ID: (Human) Q9H4X1
Entrez Gene ID: (Human) 28984
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.