Abnova
Manufacturer Code:H00028984K
Catalog # 89-200-647
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human C13ORF15 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human C13orf15 peptide using ARM Technology.
For Research Use Only
Protein Aliases: KIAA0564 MGC87338 RGC-32 RGC32 bA157L14.2; Regulator of cell cycle RGCC; Response gene to complement 32 protein; RGC-32
Gene Aliases: bA157L14.2; C13orf15; RGC-32; RGC32; RGCC
UniProt ID: (Human) Q9H4X1
Entrez Gene ID: (Human) 28984
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.