Invitrogen
Manufacturer Code:PA576411
Catalog # PIPA576411
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein of human C12orf30. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Liver regeneration-related protein LRRGT00164; Mitochondrial distribution and morphology protein 20; N-alpha-acetyltransferase 25, NatB auxiliary subunit; N-terminal acetyltransferase B complex subunit MDM20; N-terminal acetyltransferase B complex subunit NAA25; Nalpha acteyltransferase 25; NAP1 MDM20 C12orf30; NatB complex subunit MDM20
Gene Aliases: 4833422K13Rik; A630046C11; AI450868; C330023M02Rik; LRRGT00164; Mdm20; Naa25; RGD1305685
UniProt ID: (Rat) Q6QI44, (Mouse) Q8BWZ3
Entrez Gene ID: (Rat) 360811, (Mouse) 231713
Molecular Function: acetyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.