Abnova
Manufacturer Code:H00080018K
Catalog # 89-207-745
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human C12ORF30 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human C12orf30 peptide using ARM Technology.
For Research Use Only
Protein Aliases: DKFZp667K2112 FLJ13089 MDM20; mitochondrial distribution and morphology 20; Mitochondrial distribution and morphology protein 20; N-alpha-acetyltransferase 25, NatB auxiliary subunit; N-terminal acetyltransferase B complex subunit MDM20; N-terminal acetyltransferase B complex subunit NAA25; NatB complex subunit MDM20; p120
Gene Aliases: C12orf30; MDM20; NAA25; NAP1
UniProt ID: (Human) Q14CX7
Entrez Gene ID: (Human) 80018
Molecular Function: acetyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.