Proteintech
Manufacturer Code:256511AP150UL
Catalog # 50-173-5005
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | C11orf83 Fusion Protein Ag22442 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Assembly factor CBP4 homolog; C11orf83 CCDS41658.1 UNQ655 UPF0723 protein C11orf83; Ubiquinol-cytochrome-c reductase complex assembly factor 3; UPF0723 protein C11orf83
Gene Aliases: C11orf83; CCDS41658.1; MC3DN9; UNQ655; UNQ655/PRO1286; UQCC3
UniProt ID: (Human) Q6UW78
Entrez Gene ID: (Human) 790955
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.