Invitrogen
Manufacturer Code:PA595718
Catalog # PIPA595718
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 1-166 of human SDHAF2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hSDH5; SDH assembly factor 2; SDHAF2; Succinate dehydrogenase assembly factor 2, mitochondrial; succinate dehydrogenase subunit 5, mitochondrial
Gene Aliases: 0610038F07Rik; AA407634; AW049997; C11orf79; PGL2; SDH5; SDHAF2
UniProt ID: (Human) Q9NX18, (Mouse) Q8C6I2
Entrez Gene ID: (Human) 54949, (Mouse) 66072
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.