Invitrogen
Manufacturer Code:PA558904
Catalog # PIPA558904
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human C11orf79 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C11orf79 PGL2 SDH5 SDH assembly factor 2 hSDH5 succinate dehydrogenase assembly factor 2 mitochondrial succinate dehydrogenase subunit 5 mitochondrial; hSDH5; SDH assembly factor 2; SDHAF2; Succinate dehydrogenase assembly factor 2, mitochondrial; succinate dehydrogenase subunit 5, mitochondrial
Gene Aliases: C11orf79; PGL2; SDH5; SDHAF2
UniProt ID: (Human) Q9NX18
Entrez Gene ID: (Human) 54949
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.