Proteintech
Manufacturer Code:199061AP150UL
Catalog # 50-173-2593
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | C11orf79 Fusion Protein Ag13893 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C11orf79 hSDH5 PGL2 SDH assembly factor 2 SDH5 SDHAF2 SDHAF2 PGL2 SDH5; hSDH5; SDH assembly factor 2; SDH assembly factor 2 {ECO:0000255|HAMAP-Rule:MF_03057}; SDHAF2; Succinate dehydrogenase assembly factor 2, mitochondrial; succinate dehydrogenase assembly factor 2, mitochondrial {ECO:0000255|HAMAP-Rule:MF_03057}; succinate dehydrogenase subunit 5, mitochondrial; succinate dehydrogenase subunit 5, mitochondrial {ECO:0000255|HAMAP-Rule:MF_03057}
Gene Aliases: 0610038F07Rik; AA407634; AW049997; C11orf79; PGL2; RGD1309216; SDH5; SDHAF2
UniProt ID: (Human) Q9NX18, (Mouse) Q8C6I2, (Rat) Q5RJQ7
Entrez Gene ID: (Human) 54949, (Mouse) 66072, (Rat) 361726
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.