Invitrogen
Manufacturer Code:PA570253
Catalog # PIPA570253
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N-terminal region of rat Hikeshi |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Hikeshi, heat shock protein nuclear import factor; Lethal gene on chromosome 7 Rinchik 6 protein; lethal, Chr 7, Rinchik 6; Protein Hikeshi; similar to hypothetical protein HSPC138; uncharacterized protein C11orf73 homolog
Gene Aliases: 0610007P06Rik; 1110002N09Rik; C11orf73; C1H11orf73; C21H11orf73; C29H11orf73; HIKESHI; HLD13; HSPC138; HSPC179; HSPC248; l(7)6Rn; L7RN6; OPI10; zgc:110091
UniProt ID: (Bovine) Q56JY0, (Human) Q53FT3, (Zebrafish) Q568T4, (Rat) Q5M808, (Mouse) Q9DD02
Entrez Gene ID: (Bovine) 504867, (Dog) 476778, (Horse) 100050619, (Human) 51501, (Rabbit) 100343781, (Guinea pig) 100728512, (Zebrafish) 550239, (Rat) 293103, (Mouse) 67669
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.