Invitrogen
Manufacturer Code:PA557138
Catalog # PIPA557138
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human C11orf73 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HSPC179 L7RN6 lethal Chr 7 Rinchik 6 uncharacterized protein C11orf73; Lethal gene on chromosome 7 Rinchik 6 protein; lethal, Chr 7, Rinchik 6; Protein Hikeshi
Gene Aliases: 0610007P06Rik; 1110002N09Rik; C11orf73; HIKESHI; HLD13; HSPC138; HSPC179; HSPC248; l(7)6Rn; L7RN6; OPI10
UniProt ID: (Human) Q53FT3, (Rat) Q5M808, (Mouse) Q9DD02
Entrez Gene ID: (Human) 51501, (Rat) 293103, (Mouse) 67669
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.