Invitrogen
Manufacturer Code:PA524268
Catalog # PIPA524268
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 6-35 amino acids from the N-terminal region of human CK073 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: chromosome unknown open reading frame, human C11orf73; HSPC179 L7RN6 lethal Chr 7 Rinchik 6 uncharacterized protein C11orf73; lethal, Chr 7, Rinchik 6; Protein Hikeshi; Protein of unknown function DUF775 containing protein; uncharacterized protein C11orf73 homolog; Uncharacterized protein C11orf73-like
Gene Aliases: C11orf73; HIKESHI; HLD13; HSPC138; HSPC179; HSPC248; L7RN6; OPI10
UniProt ID: (Human) Q53FT3
Entrez Gene ID: (Human) 51501, (Chinese hamster) 100762248
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.