Proteintech
Manufacturer Code:204141AP150UL
Catalog # 50-173-2875
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | C11orf67 Fusion Protein Ag14258 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AAMDC C11orf67 CK067 PTD015 UPF0366 protein C11orf67; Adipogenesis associated Mth938 domain-containing protein; adipogenesis associated, Mth938 domain containing; LI2; Mth938 domain-containing protein; UPF0366 protein C11orf67; UPF0366 protein C11orf67 homolog
Gene Aliases: 1810020D17Rik; 1810037D19Rik; AAMDC; C11orf67; CK067; LI2; PTD015; RGD1561459
UniProt ID: (Human) Q9H7C9, (Mouse) Q8R0P4
Entrez Gene ID: (Human) 28971, (Mouse) 66273, (Rat) 361606
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.