Invitrogen
Manufacturer Code:PA534871
Catalog # PIPA534871
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 37 and 322 of C10orf46 (Uniprot IDNo.Q86Y37) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C10orf46 CAC1 CDK2-associated and cullin domain-containing protein 1 Cdk-Associated Cullin1; Cdk-associated cullin 1; Cdk-Associated Cullin1; CDK2-associated and cullin domain-containing protein 1; CDK2-associated, cullin domain 1; cullin
Gene Aliases: C10orf46; CAC1; CACUL1
UniProt ID: (Human) Q86Y37
Entrez Gene ID: (Human) 143384
Molecular Function: ligase ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.