Invitrogen
Manufacturer Code:MA522972
Catalog # PIMA522972
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2C4 |
Immunogen | C10orf4 (NP&lowbar660289 1 a.a. &bsim 100 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: C10orf4 F26C11.1-like FRA10A fragile site 10q23.3 protein FRA10AC1 rare folic acid-type fragile site FRA(10)(q23.3) candidate gene 1; fragile site 10q23.3; Protein FRA10AC1; Protein FRA10AC1 homolog; rare folic acid-type fragile site, FRA(10)(q23.3), candidate gene 1
Gene Aliases: C10orf4; F26C11.1-like; FRA10A; FRA10AC1; PRO2972; RGD1309482
UniProt ID: (Human) Q70Z53, (Rat) Q5FVF1
Entrez Gene ID: (Human) 118924, (Rat) 365458
Molecular Function: hydrolase phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.