Invitrogen
Manufacturer Code:PA530979
Catalog # PIPA530979
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment corresponding to a region within amino acids 1 and 192 of Human C10orf4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C10orf4 F26C11.1-like FRA10A fragile site 10q23.3 protein FRA10AC1 rare folic acid-type fragile site FRA(10)(q23.3) candidate gene 1; fragile site 10q23.3; Protein FRA10AC1; rare folic acid-type fragile site, FRA(10)(q23.3), candidate gene 1
Gene Aliases: C10orf4; F26C11.1-like; FRA10A; FRA10AC1; PRO2972
UniProt ID: (Human) Q70Z53
Entrez Gene ID: (Human) 118924
Molecular Function: hydrolase phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.