Invitrogen
Manufacturer Code:PA570010
Catalog # PIPA570010
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human C10orf132 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GOLGA7B C10orf132 C10orf133; golgi autoantigen, golgin subfamily a, 7B; golgin A7 family, member B; Golgin subfamily A member 7B
Gene Aliases: 4933417O08Rik; AI839934; bA451M19.3; bA459F3.4; C10orf132; C10orf133; GOLGA7B
UniProt ID: (Human) Q2TAP0, (Mouse) Q9D428
Entrez Gene ID: (Bovine) 767832, (Human) 401647, (Rabbit) 100356390, (Pig) 102162220, (Horse) 100070662, (Dog) 111092945, (Mouse) 71146, (Rat) 309378
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.