Invitrogen
Manufacturer Code:PA585595
Catalog # PIPA585595
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human insulin. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Insulin; Insulin A chain; Insulin B chain; Insulin-1; Insulin-1 A chain; Insulin-1 B chain; IRDN Antibody MODY10 Antibody IDDM2 Antibody INS Antibody ILPR Antibody insulin Antibody; preproinsulin; proinsulin
Gene Aliases: IDDM; IDDM1; IDDM2; ILPR; INS; Ins-1; Ins1; Ins2-rs1; IRDN; MODY10
UniProt ID: (Human) P01308, (Rat) P01322, (Mouse) P01325
Entrez Gene ID: (Human) 3630, (Rat) 24505, (Mouse) 16333
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.