OriGene
Manufacturer Code:UM870005
Catalog # 50-168-5462
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | UMAB49 |
Immunogen | Full length human recombit protein of human JUN produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Activator protein 1; AH119; AP-1 AP1 c-Jun p39; AP1; Avian sarcoma virus 17 (v-jun) oncogene homolog; enhancer-binding protein AP1; immediate early; Jun A; Jun activation domain binding protein; Jun oncogene; jun proto-oncogene; p39; Proto-oncogene c-Jun; Transcription factor AP-1; V-jun avian sarcoma virus 17 oncogene homolog; v-jun sarcoma virus 17 oncogene homolog
Gene Aliases: AP-1; AP1; c-Jun; JUN; Junc; Rjg-9
UniProt ID: (Human) P05412, (Rat) P17325, (Mouse) P05627
Entrez Gene ID: (Dog) 609429, (Human) 3725, (Rat) 24516, (Mouse) 16476
Molecular Function:
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.