Invitrogen
Manufacturer Code:PA538616
Catalog # PIPA538616
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide derived from the internal region of human CEBPE |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C/EBP delta; C/EBP epsilon; C/EBP-related protein 1; C/EBP-related protein 3; C/EBPdelta; C/EBPepsilon; CCAAT/enhancer binding protein (C/EBP), delta; CCAAT/enhancer binding protein (C/EBP), epsilon; CCAAT/enhancer binding protein , epsilon; CCAAT/enhancer binding protein epsilon; CCAAT/enhancer-binding protein delta; CCAAT/enhancer-binding protein epsilon; CEBPD CCAAT/enhancer binding protein C/EBP delta CEBPE C/EBP epsilon CCAAT/enhancer binding protein epsilon; NF-IL6-beta; Nuclear factor NF-IL6-beta; Transcription factor CELF
Gene Aliases: C/EBP-delta; C/EBP-epsilon; C/EBPd; C/EBPe; C/EPBe; CEBPD; CEBPE; CELF; CRP1; CRP3; Gm294; NF-IL6-beta
UniProt ID: (Human) P49716, (Human) Q15744, (Mouse) Q00322, (Rat) Q03484, (Rat) P56261, (Mouse) Q6PZD9
Entrez Gene ID: (Human) 1052, (Human) 1053, (Mouse) 12609, (Rat) 25695, (Rat) 25410, (Mouse) 110794
Molecular Function: nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.