Novus Biologicals
Manufacturer Code:NBP203523
Catalog # NBP203523
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1E1 |
Immunogen | Full length human recombinant protein of human BCHE (NP_000046) produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Acylcholine acylhydrolase; Acylcholine acylhydrolase Butyrylcholine esterase butyrylcholinesterase CHE1cholinesterase Choline esterase II cholinesterase 1 E1 EC 3.1.1.8 Pseudocholinesterase; Butyrylcholine esterase; Choline esterase II; Cholinesterase; cholinesterase (serum) 2; cholinesterase 1; Pseudocholinesterase
Gene Aliases: BCHE; CHE1; CHE2; E1
UniProt ID: (Human) P06276
Entrez Gene ID: (Human) 590
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.