Novus Biologicals
Manufacturer Code:NBP298274B
Catalog # NB081676
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant Butyrylcholinesterase/BCHE (NP_033868.3 Met 1-Leu 603). |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acylcholine acylhydrolase; Acylcholine acylhydrolase Butyrylcholine esterase butyrylcholinesterase CHE1cholinesterase Choline esterase II cholinesterase 1 E1 EC 3.1.1.8 Pseudocholinesterase; Butyrylcholine esterase; Choline esterase II; Cholinesterase; cholinesterase (serum) 2; cholinesterase 1; Pseudocholinesterase
Gene Aliases: BCHE; CHE1; CHE2; E1
UniProt ID: (Human) P06276
Entrez Gene ID: (Human) 590
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.