Invitrogen
Manufacturer Code:PA578861
Catalog # PIPA578861
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E.coli-derived mouse BCHE recombit protein (Position: K263-E571). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acylcholine acylhydrolase; Butyrylcholine esterase; Choline esterase II; Cholinesterase; Cholinesterase Acylcholine acylhydrolase Butyrylcholine esterase Choline esterase II Pseudocholinesterase Bche; Pseudocholinesterase
Gene Aliases: Bche; C730038G20Rik
UniProt ID: (Mouse) Q03311
Entrez Gene ID: (Mouse) 12038
Molecular Function: esterase hydrolase lipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.