Novus Biologicals
Manufacturer Code:NBP2749641.0ML
Catalog # NB029995
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | SN20-03 |
Immunogen | Recombinant protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: ATP-dependent helicase SMARCA4; ATP-dependent helicase SMARCA4 BAF190 BAF190A brahma protein-like 1 BRG1-associated factor 190A BRG1SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily A member 4 BRM/SWI2-related gene 1 EC 3.6.1 EC 3.6.4.- FLJ39786 global transcription activator homologous sequence homeotic gene regulator hSNF2b Mitotic growth and transcription activator nuclear protein GRB1 Protein brahma homolog 1 Protein BRG-1 RTPS2 SNF2 SNF2B SNF2-BETA SNF2L4 SNF2LB SNF2-like 4 subfamily a member 4 sucrose nonfermenting-like 4 SWI/SNF related matrix associated actin dependent regulator of chromatin SWI2 transcription activator BRG1; BAF190A; brahma protein-like 1; BRG1-associated factor 190A; BRM/SWI2-related gene 1; global transcription activator homologous sequence; homeotic gene regulator; Mitotic growth and transcription activator; nuclear protein GRB1; Protein brahma homolog 1; Protein BRG-1; SNF2-beta; SNF2-like 4; sucrose nonfermenting-like 4; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 4; Transcription activator BRG1
Gene Aliases: BAF190; BAF190A; BRG1; CSS4; hSNF2b; MRD16; RTPS2; SMARCA4; SNF2; SNF2B; SNF2L4; SNF2LB; SWI2
UniProt ID: (Human) P51532
Entrez Gene ID: (Human) 6597
Molecular Function:
DNA binding protein
DNA helicase
helicase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.