Invitrogen
Manufacturer Code:MA110062
Catalog # PIMA110062
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | BRG-01 |
Immunogen | Bacterially produced GST-fused protein representing 242 C-terminal amino acids of human Brg1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C do not freeze |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ATP-dependent helicase SMARCA4; BAF190 BRG1 FLJ39786 RTPS2 SNF2 SNF2-BETA SNF2L4 SNF2LB SWI2 hSNF2b ATP-dependent helicase SMARCA4 BAF190A BRG1-associated factor 190A BRM/SWI2-related gene 1 SNF2-like 4 SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 4 brahma protein-like 1 global transcription activator homologous sequence homeotic gene regulator mitotic growth and transcription activator nuclear protein GRB1 protein BRG-1 protein brahma homolog 1 sucrose nonf; BAF190A; brahma protein-like 1; BRG1-associated factor 190A; BRM/SWI2-related gene 1; global transcription activator homologous sequence; homeotic gene regulator; Mitotic growth and transcription activator; nuclear protein GRB1; Protein brahma homolog 1; Protein BRG-1; SNF2-beta; SNF2-like 4; sucrose nonfermenting-like 4; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 4; Transcription activator BRG1
Gene Aliases: BAF190; BAF190A; BRG1; CSS4; hSNF2b; MRD16; RTPS2; SMARCA4; SNF2; SNF2B; SNF2L4; SNF2LB; SWI2
UniProt ID: (Human) P51532
Entrez Gene ID: (Human) 6597
Molecular Function: DNA binding protein DNA helicase helicase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.