R&D Systems
Manufacturer Code:MAB47251100
Catalog # MAB47251100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 471916 |
Immunogen | NS0 mouse myeloma cell line transfected with human Bradykinin RB2/BDKRB2 Met1-Gln391 Accession No. P30411 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 °C as supplied. 1 month 2 to 8 °C under sterile conditions after reconstitution. 6 months -20 to -70 °C under sterile condition |
Mouse Monoclonal Antibody
Protein Aliases: B2 bradykinin receptor; B2 bradykinin receptor B2R BDKRB2 BK2 BK-2 BK-2 receptor BKR2 bradykinin receptor B2 BradykininRB2 BRB2 DKFZp686O088; B2R; BK-2 receptor
Gene Aliases: B2R; BDKRB2; BK-2; BK2; BKR2; BRB2
UniProt ID: (Human) P30411
Entrez Gene ID: (Human) 624
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.