R&D Systems
Manufacturer Code:FAB47251V100UG
Catalog # FB47251V100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 471916 |
Immunogen | NS0 mouse myeloma cell line transfected with human Bradykinin RB2/BDKRB2 Met1-Gln391 Accession # P30411 |
Conjugate | Alexa Fluor 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8°C as supplied. |
Mouse Monoclonal Antibody
Protein Aliases: B2 bradykinin receptor; B2 bradykinin receptor B2R BDKRB2 BK2 BK-2 BK-2 receptor BKR2 bradykinin receptor B2 BradykininRB2 BRB2 DKFZp686O088; B2R; BK-2 receptor
Gene Aliases: B2R; BDKRB2; BK-2; BK2; BKR2; BRB2
UniProt ID: (Human) P30411
Entrez Gene ID: (Human) 624
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.